Learn More
Medchemexpress LLC IL-20R beta Rat HE 10ug | 10ug
SDP
Catalog No. 5000185953
Encompass_Preferred
IL-20R beta Protein (IL20RB) is the beta subunit of IL-20 receptor forms functional heterodimers with different subunit protein and involves in STAT3 activation pathway[1] The IL-20RA/IL-20RB dimer is a receptor for IL-19 IL-20 and IL-24 The IL-22RA1/IL-20RB dimer is a receptor for IL-20 and IL-24[2] IL-20R beta Protein is consists of 308 amino acids (M1-T308) with a transmembrane domain (226-249 a a ) IL-20R beta Protein Rat (HEK293 Fc) is produced in HEK293 cells with a C-Terminal hFc-tag
Catalog No. 50-001-85953 Supplier Medchemexpress LLC Supplier No. HYP7700010UG
IL-20R beta Protein (IL20RB) is the beta subunit of IL-20 receptor forms functional heterodimers with different subunit protein and involves in STAT3 activation pathway[1] The IL-20RA/IL-20RB dimer is a receptor for IL-19 IL-20 and IL-24 The IL-22RA1/IL-20RB dimer is a receptor for IL-20 and IL-24[2] IL-20R beta Protein is consists of 308 amino acids (M1-T308) with a transmembrane domain (226-249 a a ) IL-20R beta Protein Rat (HEK293 Fc) is produced in HEK293 cells with a C-Terminal hFc-tag