Learn More
ACROBiosystems ACROBiosystems Human IL-21 Protein, Fc Tag
Supplier:  ACROBiosystems IL1H5253100UG
Encompass_Preferred
Recombinant Protein;100UG;Human IL-21, Fc Tag (IL1-H5253) is expressed from human 293 cells (HEK293). It contains AA Gln 23 - Ser 155 (Accession # Q9HBE4-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 41.9 kDa. The protein migrates as 45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human IL-21 R, His Tag (Cat. No. ILR-H5226) at 2 μg/mL (100 μL/well) can bind Human IL-21, Fc Tag (Cat. No. IL1-H5253) with a linear range of 0.2-8 ng/mL (QC tested).
Catalog No. 50-195-7131