Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC IL-21 protein, human | >95.0% | 100UG
SDP

Supplier:  Medchemexpress LLC HYP7038100UG

Encompass_Preferred

Recombinant human interleukin-21 provided as a lyophilized purified protein for research use in immunology and cell signaling applications. The material is expressed in E. coli, has an apparent molecular weight of ~15.4-18 kDa, and is characterized by >95% purity by reducing SDS-PAGE. Formulation options and storage guidance are provided to support stability and downstream assays.

  • Lyophilized recombinant human IL-21 suitable for biological assays.
  • Expressed in E. coli and purified to >95% purity by reducing SDS-PAGE.
  • Apparent molecular weight ~15.4-18 kDa.
  • Formulated after dialysis against PBS or phosphate buffer with stabilizers such as trehalose, mannitol, and Tween-80.
  • Stable frozen; store lyophilized at -20°C and aliquot for long-term storage.
  • Intended for immunology assays involving T cells, B cells, and NK cells.

Catalog No. 50-001-76678


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.