Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Recombinant human interleukin-21 (rHu IL-21) | >95.0% | 2UG
SDP

Catalog No. 5000176123
Encompass_Preferred

Recombinant human interleukin-21 (rHu IL-21) is a tag-free, lyophilized cytokine produced in E. coli representing residues Q32-S162 of the human protein. It is provided with >95% purity and low endotoxin, and is intended for receptor binding studies, cell-based assays, and functional activity testing.

  • Tag-free recombinant human sequence (Q32-S162).
  • Lyophilized powder for convenient storage and handling.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Low endotoxin level (<1 EU/μg).
  • Predicted molecular mass 15.6 kDa; observed ~15.4-18.0 kDa by SDS-PAGE.
  • Demonstrated biological activity with ED50 <0.5 ng/mL in cell assays.

Catalog No. 50-001-76123 Supplier Medchemexpress LLC Supplier No. HYP70382UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human interleukin-21 (rHu IL-21) is a tag-free, lyophilized cytokine produced in E. coli representing residues Q32-S162 of the human protein. It is provided with >95% purity and low endotoxin, and is intended for receptor binding studies, cell-based assays, and functional activity testing.

  • Tag-free recombinant human sequence (Q32-S162).
  • Lyophilized powder for convenient storage and handling.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Low endotoxin level (<1 EU/μg).
  • Predicted molecular mass 15.6 kDa; observed ~15.4-18.0 kDa by SDS-PAGE.
  • Demonstrated biological activity with ED50 <0.5 ng/mL in cell assays.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.