Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human IL-21 ELISA Kit

Catalog No. 5017939
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-179-39 96 Tests
BMS2043TEN 10 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 50-179-39 Supplier Invitrogen™ Supplier No. BMS2043
Add to Cart
Add to Cart
This item is not returnable. View return policy

Includes

Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human IL-8/NAP-1
Biotin-Conjugate anti-human IL-8/NAP-1 polyclonal antibody
Streptavidin-HRP
Human IL-8/NAP-1 Standard, 100ng/mL
Control high, lyophilized
Control low, lyophilized
Sample Diluent
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Red-Dye
Adhesive Films

ELISA

The Human Interleukin-8 (Neutrophil Activating Peptide-1) (Hu IL-8) ELISA quantitates Hu IL-8 in human serum, plasma, amniotic fluid, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu IL-8. Principle of the method The Human IL-8 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

IL-21 is a 17 kDa immunomodulatory cytokine produced mainly by Natural Killer Cells (NKT), T helper (Th) 17 and T follicular helper (TFH) cells. In TFH cells, IL-21 expression leads to autocrine signaling through the IL-21 receptor (IL-21R) and STAT3, which leads to additional transcriptional activation by Bcl6. As with IFN gamma for Th1 and IL-4 for Th2 cells, IL-21 is critical for TFH cell development and effector function. IL-21 plays a role in T cell-dependent B cell differentiation into plasma cells and memory cells, stimulation of IgG production and induction of apoptotic signaling in naive B cells. In Th17 cells, IL-21 expression and autocrine feedback through STAT3, IRF4 and ROR gamma t lead to upregulation of the IL-23R, thereby preparing Th17 cells for maturation and maintenance by the inflammatory cytokine IL-23. While upregulating IRF4 and ROR gamma t, IL-21 also mediates the downregulation of Foxp3. IL-21 deficient mice are protected from developing colitis upon chemical treatment by their inability to upregulate Th17-associated molecules. In comparison, elevated levels of IL-21 are present in chemically-induced colitis mouse models.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9HBE4
Assay Range 78-5000 pg/mL
Assay Sensitivity 20.0 pg/mL
Content And Storage Aluminum pouch(es) with a Microplate coated with monoclonal antibody to human IL-21
Biotin-Conjugate anti-human IL-21 monoclonal antibody
Streptavidin-HRP
Human IL-21 Standard lyophilized, 10 ng/mL upon reconstitution
Sample Diluent
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Adhesive Films
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
Target Species Human
For Use With (Application) ELISA
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias CVID11; CX2; il 21; IL21; IL-21; ILN; Interleukin; interleukin 21; Interleukin21; interleukin-21; interleukin-21 isoform; porcine interleukin-21; Za11CVID11; CX2; il 21; IL21; IL-21; ILN; Interleukin; interleukin 21; Interleukin21; interleukin-21; interleukin-21 isoform; porcine interleukin-21; Za11; CVID11; IL-21; Za11
Gene ID (Entrez) 59067
Gene Symbol IL21
Interassay CV 7.7%
Intraassay CV 5.8%
Kit Contents Pre-coated 96 well plate, Standard, Sample Diluent, Assay Buffer concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Sample Diluent, Assay Buffer concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Protein Aliases IL-21, Za11
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2-8°C
Synonym Protein 3-10 C, CXCL8, NAP-1, GCP-1
Target IL-21
Test Time 1 hr 20 min
Total Assay Time 4 hr 10 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.