Learn More
Medchemexpress LLC IL-21 Human His-Av 20ug | 20ug
SDP
Catalog No. 5000193940
Encompass_Preferred
IL-21 protein is an immunomodulatory cytokine that promotes the transition from innate immunity to adaptive immunity It induces B cells to produce IgG(1) and IgG(3) harnessing an effective antibody response to fight viral infections IL-21 Protein Human (His-Avi) is the recombinant human-derived IL-21 protein expressed by E coli with N-His N-Avi labeled tag
Catalog No. 50-001-93940 Supplier Medchemexpress LLC Supplier No. HYP7814820UG
IL-21 protein is an immunomodulatory cytokine that promotes the transition from innate immunity to adaptive immunity It induces B cells to produce IgG(1) and IgG(3) harnessing an effective antibody response to fight viral infections IL-21 Protein Human (His-Avi) is the recombinant human-derived IL-21 protein expressed by E coli with N-His N-Avi labeled tag