Learn More
Medchemexpress LLC HY-P7078 10ug Medchemexpress, IL-21, Mouse CAS: Purity:>98%
SDP
Catalog No. 501934320
Encompass_Preferred
Medchemexpress, HY-P7078 10ug IL-21, Mouse CAS: Recombinant Mouse Interleukin-21 is a regulatory cytokine thought to play a role in the transition between innate and adaptive immunity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4320 Supplier Medchemexpress LLC Supplier No. HYP707810UG
Medchemexpress, HY-P7078 10ug IL-21, Mouse CAS: Recombinant Mouse Interleukin-21 is a regulatory cytokine thought to play a role in the transition between innate and adaptive immunity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.