Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC IL-21 Mouse 129a.a 100ug
SDP

Catalog No. 5000196991
Encompass_Preferred

Recombinant mouse interleukin-21 (129 amino acids) supplied as a lyophilized protein for research use in immunology and cell signaling studies. The material corresponds to the mature mouse IL-21 sequence (H18-S146; accession Q9ES17), is expressed in Escherichia coli, and is provided with specifications for purity and endotoxin suitable for in vitro assays.

  • Sequence corresponds to mature mouse IL-21 (129 a.a.; H18-S146).
  • Expressed in Escherichia coli and supplied lyophilized for storage stability.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Endotoxin level below 0.2 EU/μg for sensitive assays.
  • Approximate molecular weight ~15.1 kDa under reducing conditions.
  • Recommended storage at -20°C; aliquot and avoid repeated freeze-thaw.

Catalog No. 50-001-96991 Supplier Medchemexpress LLC Supplier No. HYP7078100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant mouse interleukin-21 (129 amino acids) supplied as a lyophilized protein for research use in immunology and cell signaling studies. The material corresponds to the mature mouse IL-21 sequence (H18-S146; accession Q9ES17), is expressed in Escherichia coli, and is provided with specifications for purity and endotoxin suitable for in vitro assays.

  • Sequence corresponds to mature mouse IL-21 (129 a.a.; H18-S146).
  • Expressed in Escherichia coli and supplied lyophilized for storage stability.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Endotoxin level below 0.2 EU/μg for sensitive assays.
  • Approximate molecular weight ~15.1 kDa under reducing conditions.
  • Recommended storage at -20°C; aliquot and avoid repeated freeze-thaw.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.