Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC IL-21 protein, canine | >95.0% | 100 UG
SDP

Catalog No. 5000293478
Encompass_Preferred

Recombinant canine interleukin-21 (IL-21) protein expressed in E. coli and supplied as a lyophilized, tag-free preparation for immunology research. IL-21 modulates B-cell and T-cell responses, promotes NK-cell proliferation, and is suitable for assay development and cytokine signaling studies.

  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Tag-free recombinant protein expressed in E. coli.
  • Lyophilized formulation; follow datasheet reconstitution instructions.
  • Stable for long-term storage at -20°C; aliquot and store at -80°C for extended stability.
  • Sequence corresponds to accession Q6L7I9 (H18-S146).
  • Supplied as 100 μg of material suitable for research and assay use.

Catalog No. 50-002-93478 Supplier Medchemexpress LLC Supplier No. HYP74819100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant canine interleukin-21 (IL-21) protein expressed in E. coli and supplied as a lyophilized, tag-free preparation for immunology research. IL-21 modulates B-cell and T-cell responses, promotes NK-cell proliferation, and is suitable for assay development and cytokine signaling studies.

  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Tag-free recombinant protein expressed in E. coli.
  • Lyophilized formulation; follow datasheet reconstitution instructions.
  • Stable for long-term storage at -20°C; aliquot and store at -80°C for extended stability.
  • Sequence corresponds to accession Q6L7I9 (H18-S146).
  • Supplied as 100 μg of material suitable for research and assay use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.