Learn More
ACROBiosystems IL-21 R IL-2 R gamma 100ug
Catalog No. 502534604
Encompass_Preferred
Human IL-21 R IL-2 R gamma Heterodimer Protein Fc TagFc Tag (ILA-H52W9) is expressed from human 293 cells (HEK293). It contains AA Cys 20 - Pro 236 (IL-21 R) Leu 23 - Asn 254 (IL-2 R gamma) (Accession # Q9HBE5-1 (IL-21 R) P31785-1 (IL-2 R gamma)).
Catalog No. 50-253-4604 Supplier ACROBiosystems Supplier No. ILAH52W9100UG
Human IL-21 R IL-2 R gamma Heterodimer Protein Fc TagFc Tag (ILA-H52W9) is expressed from human 293 cells (HEK293). It contains AA Cys 20 - Pro 236 (IL-21 R) Leu 23 - Asn 254 (IL-2 R gamma) (Accession # Q9HBE5-1 (IL-21 R) P31785-1 (IL-2 R gamma)).