Learn More
Cell Sciences Inc IL-21 RECOMBINANT HUMAN 10UG
Supplier:  Cell Sciences Inc CRI172B
Encompass
Recombinant Human Interleukin-21 is a single, non-glycosylated polypeptide chain containing 133 amino acids. Made in E. coli. MW 15.4 kDa. > 97% pure by SDS-PAGE & HPLC. Lyophilized.
Catalog No. NC0262203