Learn More
Enzo Life Sciences IL-21R (ED) polyclonal antibody (100µg)
SDP
Supplier:  Enzo Life Sciences ADI905308100
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to a portion of human IL-21R precursor. Species reactivity: Human, Mouse, Rat. Applications: IHC, WB. Purity: Peptide affinity purified. Formulation: Liquid. In PBS containing 0.02% sodium azide.
Catalog No. 50-200-6964