Learn More
Medchemexpress LLC IL-21R Rat HEK293 2ug
SDP
Catalog No. 5000181318
Encompass_Preferred
IL-21R Protein a receptor for interleukin-21 is a type I cytokine receptor It forms a heterodimer with the common gamma subunit and associates with Janus kinase 1 (JAK1) These interactions are pivotal for transducing the cellular responses initiated by interleukin-21 regulating immune processes and inflammatory pathways IL-21R Protein Rat (HEK293 His) is the recombinant rat-derived IL-21R protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-81318 Supplier Medchemexpress LLC Supplier No. HYP731912UG
IL-21R Protein a receptor for interleukin-21 is a type I cytokine receptor It forms a heterodimer with the common gamma subunit and associates with Janus kinase 1 (JAK1) These interactions are pivotal for transducing the cellular responses initiated by interleukin-21 regulating immune processes and inflammatory pathways IL-21R Protein Rat (HEK293 His) is the recombinant rat-derived IL-21R protein expressed by HEK293 with C-His labeled tag