Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC IL-23 Human HEK293 100ug
SDP

Supplier:  Medchemexpress LLC HYP70736100UG

Encompass_Preferred

Recombinant human IL-23 protein is a heterodimeric cytokine composed of IL-23 alpha (p19) and IL-12 beta (p40), expressed in HEK293 cells and supplied as a C-terminal 6xHis-tagged, lyophilized product for research use. It is suitable for studies of IL-23 signaling, Th17 biology, and inflammatory pathways, and is provided with high purity and recommended storage conditions for long-term stability.

  • Contains the biologically relevant p19/p40 heterodimer.
  • Expressed in HEK293 mammalian cells for native-like post-translational modifications.
  • Supplied as a lyophilized powder for long-term storage.
  • Purity greater than 95% by reducing SDS-PAGE.
  • C-terminal 6xHis tag facilitates detection and purification.
  • Stable when aliquoted and stored at -20°C or -80°C.

Catalog No. 50-001-97062


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.