Learn More
Medchemexpress LLC IL-23 Human HEK293 100ug
SDP
Catalog No. 5000182366
Encompass_Preferred
IL-23 and IL12B together constitute the pro-inflammatory cytokine IL-23 which is crucial in innate and adaptive immunity IL-23 is released by antigen-presenting cells such as dendritic cells or macrophages binds to IL12RB1 and IL23R and activates JAK2 and TYK2 IL-23 Protein Human (HEK293 His) is a recombinant protein dimer complex containing human-derived IL-23 alpha IL-12 beta expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-82366 Supplier Medchemexpress LLC Supplier No. HYP73193100UG
IL-23 and IL12B together constitute the pro-inflammatory cytokine IL-23 which is crucial in innate and adaptive immunity IL-23 is released by antigen-presenting cells such as dendritic cells or macrophages binds to IL12RB1 and IL23R and activates JAK2 and TYK2 IL-23 Protein Human (HEK293 His) is a recombinant protein dimer complex containing human-derived IL-23 alpha IL-12 beta expressed by HEK293 with C-His labeled tag