Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC IL-23 Human HEK293 10ug
SDP

Catalog No. 5000185222
Encompass_Preferred

Recombinant human IL-23 is a heterodimeric cytokine composed of IL-23 alpha and IL-12 beta subunits, expressed in HEK293 cells and supplied lyophilized for research applications. It is suitable for studies of IL-23 receptor interaction and downstream JAK2/TYK2 signaling in immunology and inflammation; refer to the lot-specific COA for buffer and bioactivity details.

  • Contains IL-23 alpha and IL-12 beta heterodimer.
  • Expressed in mammalian HEK293 cells for native glycosylation.
  • Supplied lyophilized for extended shelf life.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Stable when stored at -20°C; aliquot and freeze for long-term storage.
  • Recommended reconstitution concentration is at least 100 μg/mL in ddH2O; consult COA for buffer specifics.

Catalog No. 50-001-85222 Supplier Medchemexpress LLC Supplier No. HYP7073610UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human IL-23 is a heterodimeric cytokine composed of IL-23 alpha and IL-12 beta subunits, expressed in HEK293 cells and supplied lyophilized for research applications. It is suitable for studies of IL-23 receptor interaction and downstream JAK2/TYK2 signaling in immunology and inflammation; refer to the lot-specific COA for buffer and bioactivity details.

  • Contains IL-23 alpha and IL-12 beta heterodimer.
  • Expressed in mammalian HEK293 cells for native glycosylation.
  • Supplied lyophilized for extended shelf life.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Stable when stored at -20°C; aliquot and freeze for long-term storage.
  • Recommended reconstitution concentration is at least 100 μg/mL in ddH2O; consult COA for buffer specifics.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.