Learn More
Medchemexpress LLC IL-23 Human HEK293 2ug
SDP
Supplier:  Medchemexpress LLC HYP707362UG
Encompass_Preferred
Recombinant human interleukin-23 (IL-23) is supplied as a heterodimeric protein expressed in HEK293 cells with a C-terminal 6xHis tag and provided lyophilized for research use. It is suitable for in vitro assays examining IL-23 signaling, receptor binding, and related immunological studies.
  • Human sequence expressed in HEK293 to retain mammalian post-translational modifications.
  • C-terminal 6xHis tag for straightforward detection and purification.
  • Lyophilized formulation for extended shelf life and convenient storage.
  • High purity (>95%) for reproducible experimental results.
  • Small 2 μg pack size suitable for assay development and optimization.
Catalog No. 50-001-96981