Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC IL-23 Mouse HEK293 10ug
SDP

Catalog No. 5000184714
Encompass_Preferred

Recombinant mouse IL-23 is a heterodimeric cytokine consisting of IL-23 alpha (p19) and IL-12 beta (p40) subunits, produced in HEK293 cells and supplied lyophilized for research applications. The protein is tag free, >95% pure by reducing SDS-PAGE, formulated from 0.2 μm filtered PBS (pH 7.4), and should be aliquoted and stored at -20 °C or -80 °C for extended storage.

  • Expressed in HEK293 cells.
  • Tag free heterodimeric protein.
  • Lyophilized from PBS, pH 7.4.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Recommended storage at -20 °C or -80 °C for extended storage.
  • Available in 2 μg, 10 μg, 50 μg, and 100 μg sizes.

Catalog No. 50-001-84714 Supplier Medchemexpress LLC Supplier No. HYP7070910UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant mouse IL-23 is a heterodimeric cytokine consisting of IL-23 alpha (p19) and IL-12 beta (p40) subunits, produced in HEK293 cells and supplied lyophilized for research applications. The protein is tag free, >95% pure by reducing SDS-PAGE, formulated from 0.2 μm filtered PBS (pH 7.4), and should be aliquoted and stored at -20 °C or -80 °C for extended storage.

  • Expressed in HEK293 cells.
  • Tag free heterodimeric protein.
  • Lyophilized from PBS, pH 7.4.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Recommended storage at -20 °C or -80 °C for extended storage.
  • Available in 2 μg, 10 μg, 50 μg, and 100 μg sizes.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.