Learn More
Sino Biological Recombinant Human IL-23 Protein (His Tag)
Supplier:  Sino Biological CT012H08H1
Encompass_Preferred
A DNA sequence encoding the IL23A (NP_057668.1) (Met1-Pro189) was fused with a polyhistidine tag at the C-terminus, constructed the plasmid 1; A DNA sequence encoding the p40 subunit of human IL12, termed as IL12B (NP_002178.2) (Met1-Ser328) was fused with a polyhistidine tag at the C-terminus, constructed the plasmid 2. The two plasmids were co-expressed and the IL23 heterodimer was purified.
Catalog No. 50-211-6320