Learn More
BPS Bioscience Inc IL-23 Responsive STAT3 Luciferase Reporter HEK293 Cell Line, 2 vials, >1 x 10^6 cells per vial, BSL-2, -80°C storage
SDP
Supplier:  BPS Bioscience Inc 82591
Encompass
IL-23 Responsive STAT3 Luciferase Reporter HEK293 Cell Line, 2 vials, >1 x 10^6 cells per vial, BSL-2, -80°C storage
Catalog No. NC3594644
Add to Cart
Request a Quote