Learn More
AdipoGen anti-IL-23p19 (human), mAb (I 178G) (50 µg)
SDP
Supplier:  AdipoGen AG20A0027C050
Encompass_Preferred
Monoclonal Antibody. Recognizes human IL-23p19. Isotype: Rat IgG2akappa. Clone: I 178G. Applications: ELISA, WB. Liquid. 0.2µm-filtered solution in PBS, pH 7.4. Contains no preservatives. IL-23p19 associates with IL-12b to form the IL-23 interleukin, an heterodimeric cytokine which functions in innate and adaptive immunity. IL-23 binds to an heterodimeric receptor complex composed of IL-12RB1 and IL-23R, activates the Jak-Stat signaling cascade, stimulates memory rather than naive T-cells and promotes production of proinflammatory cytokines. IL-23 induces autoimmune inflammation and thus may be responsible for autoimmune inflammatory diseases and may be important for tumorigenesis.
Catalog No. 50-205-2807