Learn More
ACROBiosystems ACROBiosystems Human IL-23 R Protein, His Tag
Supplier:  ACROBiosystems ILRH52H41MG
Encompass_Preferred
Recombinant Protein;1MG;Human IL-23 R, His Tag (ILR-H52H4) is expressed from human 293 cells (HEK293). It contains AA Gly 24 - Gly 355 (Accession # Q5VWK5-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 39.9 kDa. The protein migrates as 55-80 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Captured Biotinylated Human IL23A & IL12B, His Tag, Avi Tag (Cat. No. ILB-H82W6) on Biotin CAP - Series S sensor Chip can bind Human IL-23 R, His Tag (Cat. No. ILR-H52H4) affinity constant of 4.77 nM as determined in a SPR assay (Biacore T200) (Routinely tested).
Catalog No. 50-195-7150