Learn More
Medchemexpress LLC IL-23R Rat HEK293 100ug
SDP
Catalog No. 5000182290
Encompass_Preferred
IL-23R Protein critical in immune response forms the interleukin-23 receptor by associating with IL12RB1 Binding to IL23 it activates T-cells NK cells and potentially macrophage/myeloid cells through Jak-Stat signaling Associated with autoimmune diseases and tumorigenesis IL-23R in the presence of IL12RB1 forms a heterodimeric IL-23 receptor interacting with JAK2 and activating STAT3 IL-23R Protein Rat (HEK293 Fc) is the recombinant rat-derived IL-23R protein expressed by HEK293 with C-hFc labeled tag
Catalog No. 50-001-82290 Supplier Medchemexpress LLC Supplier No. HYP73197100UG
IL-23R Protein critical in immune response forms the interleukin-23 receptor by associating with IL12RB1 Binding to IL23 it activates T-cells NK cells and potentially macrophage/myeloid cells through Jak-Stat signaling Associated with autoimmune diseases and tumorigenesis IL-23R in the presence of IL12RB1 forms a heterodimeric IL-23 receptor interacting with JAK2 and activating STAT3 IL-23R Protein Rat (HEK293 Fc) is the recombinant rat-derived IL-23R protein expressed by HEK293 with C-hFc labeled tag