Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Interleukin-23 receptor protein, rat (HEK293, hFc) | >95.0% | 50UG
SDP

Catalog No. 5000175248
Encompass_Preferred

Recombinant rat interleukin-23 receptor fragment expressed in HEK293 cells with a C-terminal human Fc tag. Supplied lyophilized from PBS with trehalose, the protein is designed for biochemical and biophysical assays and is suitable for binding studies such as surface plasmon resonance to characterize IL-23/IL-23R interactions.

  • Recombinant rat IL-23R (G39-D367) with C-terminal hFc tag.
  • Expressed in HEK293 mammalian cells for native glycosylation.
  • Purity greater than 95% as determined by Bis-Tris PAGE.
  • Predicted molecular weight 63.61 kDa; observed 90-120 kDa on SDS-PAGE.
  • Lyophilized formulation in PBS, pH 7.4, with 8% trehalose.
  • Validated for surface plasmon resonance (SPR) binding studies.
  • Store at -20°C; freeze aliquots at -20°C or -80°C for extended storage.

Catalog No. 50-001-75248 Supplier Medchemexpress LLC Supplier No. HYP70367850UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Recombinant rat interleukin-23 receptor fragment expressed in HEK293 cells with a C-terminal human Fc tag. Supplied lyophilized from PBS with trehalose, the protein is designed for biochemical and biophysical assays and is suitable for binding studies such as surface plasmon resonance to characterize IL-23/IL-23R interactions.

  • Recombinant rat IL-23R (G39-D367) with C-terminal hFc tag.
  • Expressed in HEK293 mammalian cells for native glycosylation.
  • Purity greater than 95% as determined by Bis-Tris PAGE.
  • Predicted molecular weight 63.61 kDa; observed 90-120 kDa on SDS-PAGE.
  • Lyophilized formulation in PBS, pH 7.4, with 8% trehalose.
  • Validated for surface plasmon resonance (SPR) binding studies.
  • Store at -20°C; freeze aliquots at -20°C or -80°C for extended storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.