Learn More
Medchemexpress LLC IL-27 Human CHO H 10ug
SDP
Catalog No. 5000181156
Encompass_Preferred
IL-27 (Interleukin-27) is a heterodimeric cytokine of the IL-12 family composed of two subunits EBI3 (Epstein-Barr-virus-induced molecule 3) and p28[1] IL-27 enhances the development proliferation and cytotoxic activity of CD8 T cells thereby indirectly promoting anti-tumor immunity[ 2] IL-27 Protein Human (CHO His) is a recombinant protein with a His label that consists of two subunits EBI3 (IL27B) (229 amino acids (M1-K229)) and IL27A (214 amino acids (F29-P243)) is produced by CHO cells
Catalog No. 50-001-81156 Supplier Medchemexpress LLC Supplier No. HYP7319910UG
IL-27 (Interleukin-27) is a heterodimeric cytokine of the IL-12 family composed of two subunits EBI3 (Epstein-Barr-virus-induced molecule 3) and p28[1] IL-27 enhances the development proliferation and cytotoxic activity of CD8 T cells thereby indirectly promoting anti-tumor immunity[ 2] IL-27 Protein Human (CHO His) is a recombinant protein with a His label that consists of two subunits EBI3 (IL27B) (229 amino acids (M1-K229)) and IL27A (214 amino acids (F29-P243)) is produced by CHO cells