Learn More
Medchemexpress LLC IL-27 Mouse HEK293 10ug
SDP
Catalog No. 5000181316
Encompass_Preferred
The IL-27 protein together with IL23A forms IL-23 interleukin a key cytokine in innate and adaptive immunity It is associated with infection responses binds to IL12RB1 and IL23R receptor complexes and activates Jak-Stat signaling IL-27 Protein Mouse (HEK293 C-His) is a recombinant protein dimer complex containing IL-27 Protein and IL-27A expressed by HEK293 with C-10 His labeled tag and GSGSGGSGGSGSGKL peptide
Catalog No. 50-001-81316 Supplier Medchemexpress LLC Supplier No. HYP73200A10UG
The IL-27 protein together with IL23A forms IL-23 interleukin a key cytokine in innate and adaptive immunity It is associated with infection responses binds to IL12RB1 and IL23R receptor complexes and activates Jak-Stat signaling IL-27 Protein Mouse (HEK293 C-His) is a recombinant protein dimer complex containing IL-27 Protein and IL-27A expressed by HEK293 with C-10 His labeled tag and GSGSGGSGGSGSGKL peptide