Learn More
Sino Biological Recombinant Human IL-27 Protein (His Tag)
Supplier:  Sino Biological 10076H08S100
Encompass_Preferred
A DNA sequence encoding the Human IL27 complex composed of IL27B subunit (NP_005746.2) (Met 1-Lys 229) and the N-terminally truncated IL27p28 (IL27A) subunit (NP_663634.2) (Phe 29-Pro 243) linked by an polypeptide linker was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-211-6269