Learn More
ACROBiosystems ACROBiosystems Human IL-27 Ra / TCCR Protein, Fc Tag
Supplier:  ACROBiosystems ILAH52541MG
Encompass_Preferred
Recombinant Protein;1MG;Human IL-27 Ra, Fc Tag (ILA-H5254) is expressed from human 293 cells (HEK293). It contains AA Gln 33 - Arg 514 (Accession # AAH28003).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 78.9 kDa. The protein migrates as 105-115 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-195-7103