Learn More
Medchemexpress LLC IL-2R beta/CD122 Cy 100ug | 100ug
SDP
Catalog No. 5000187599
Encompass_Preferred
IL-2R beta (CD122) a type cytokine receptor expressed on T lymphocytes is a receptor for IL-2 (Kd 1 nM approximately)[1] IL-2R beta mediates T cell immune responses such as stimulating T cell proliferation and activating lymphokine-activated killer cells[2] IL-2R beta/CD122 Protein Cynomolgus (HEK293 His) is a recombinant cynomolgus IL-2R beta (A27-D239) with a C-Terminal His tag which is produced in HEK 293
Catalog No. 50-001-87599 Supplier Medchemexpress LLC Supplier No. HYP77883100UG
IL-2R beta (CD122) a type cytokine receptor expressed on T lymphocytes is a receptor for IL-2 (Kd 1 nM approximately)[1] IL-2R beta mediates T cell immune responses such as stimulating T cell proliferation and activating lymphokine-activated killer cells[2] IL-2R beta/CD122 Protein Cynomolgus (HEK293 His) is a recombinant cynomolgus IL-2R beta (A27-D239) with a C-Terminal His tag which is produced in HEK 293