Learn More
Medchemexpress LLC IL-2R beta/CD122 Cy 20ug | 20ug
SDP
Catalog No. 5000187657
Encompass_Preferred
IL-2R beta (CD122) a type cytokine receptor expressed on T lymphocytes is a receptor for IL-2 (Kd 1 nM approximately)[1] IL-2R beta mediates T cell immune responses such as stimulating T cell proliferation and activating lymphokine-activated killer cells[2] IL-2R beta/CD122 Protein Cynomolgus (HEK293 His) is a recombinant cynomolgus IL-2R beta (A27-D239) with a C-Terminal His tag which is produced in HEK 293
Catalog No. 50-001-87657 Supplier Medchemexpress LLC Supplier No. HYP7788320UG
IL-2R beta (CD122) a type cytokine receptor expressed on T lymphocytes is a receptor for IL-2 (Kd 1 nM approximately)[1] IL-2R beta mediates T cell immune responses such as stimulating T cell proliferation and activating lymphokine-activated killer cells[2] IL-2R beta/CD122 Protein Cynomolgus (HEK293 His) is a recombinant cynomolgus IL-2R beta (A27-D239) with a C-Terminal His tag which is produced in HEK 293