Learn More
Medchemexpress LLC HY-P7374 10ug Medchemexpress, IL-2R α/CD25, Fc Chimera (HEK293-expressed), Human CAS: Purity:>98%
SDP
Catalog No. 502083155
Encompass_Preferred
Medchemexpress, HY-P7374 10ug IL-2R α/CD25, Fc Chimera (HEK293-expressed), Human CAS: Recombinant Human Interleukin-2 Receptor alpha/CD25, Fc Chimera (HEK293-expressed) is a subunit of interleukin-2 receptor, plays a critical role in regulating the IL-2-mediated responses by modulating the affinity of the IL-2R for the ligand. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3155 Supplier Medchemexpress LLC Supplier No. HYP737410UG
Medchemexpress, HY-P7374 10ug IL-2R α/CD25, Fc Chimera (HEK293-expressed), Human CAS: Recombinant Human Interleukin-2 Receptor alpha/CD25, Fc Chimera (HEK293-expressed) is a subunit of interleukin-2 receptor, plays a critical role in regulating the IL-2-mediated responses by modulating the affinity of the IL-2R for the ligand. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.