Learn More
Enzo Life Sciences IL-2Ralpha (CD25) Recombinant monoclonal antibody (Human IgG1 kappa) (200µg)
SDP
Supplier:  Enzo Life Sciences ENZABS5450200
Encompass_Preferred
Clone: Daclizumab. Isotype: IgG1κ. Immunogen: Cultured T-cells derived from a patient with Mycosis fungoides. Species reactivity: Human. Specificity: Recognizes the p55 chain of IL2R on human T cells, Tac antigen, which is expressed on T cells activated by mitogens, soluble antigens, and alloantigens but not on resting T cells, thymocytes, B cells, monocytes, activated B cells, leukemic T cell blasts from patients with acute lymphoblastic leukemia, and long-term T cell lines. Applications: Flow Cytometry, FUNC. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 0.02% Proclin 300.
Catalog No. 50-201-3071