Learn More
ACROBiosystems ACROBiosystems Biotinylated Human IL-2 R alpha / CD25 Protein, His,Avitag™
Catalog No. 501956970
Encompass_Preferred
Recombinant Protein;200UG;Biotinylated Human IL-2 R alpha, His,Avitag (ILA-H82E6) is expressed from human 293 cells (HEK293). It contains AA Glu 22 - Cys 213 (Accession # P01589-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 25.4 kDa. The protein migrates as 35-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human IL-2, Tag Free (Cat. No. IL2-H4113) at 5 μg/mL (100 μL/well) can bind Biotinylated Human IL-2 R alpha, His,Avitag (Cat. No. ILA-H82E6) with a linear range of 10-156 ng/mL (QC tested).;Biotinylated Human IL-2 R alpha, His,Avitag (Cat. No. ILA-H82E6) inhibits the IL-2-dependent proliferation of Mo7e cells. The EC50 for this effect is 0.57-0.81 µg/mL (Routinely tested).
Catalog No. 50-195-6970 Supplier ACROBiosystems Supplier No. ILAH82E6200UG
Recombinant Protein;200UG;Biotinylated Human IL-2 R alpha, His,Avitag (ILA-H82E6) is expressed from human 293 cells (HEK293). It contains AA Glu 22 - Cys 213 (Accession # P01589-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 25.4 kDa. The protein migrates as 35-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human IL-2, Tag Free (Cat. No. IL2-H4113) at 5 μg/mL (100 μL/well) can bind Biotinylated Human IL-2 R alpha, His,Avitag (Cat. No. ILA-H82E6) with a linear range of 10-156 ng/mL (QC tested).;Biotinylated Human IL-2 R alpha, His,Avitag (Cat. No. ILA-H82E6) inhibits the IL-2-dependent proliferation of Mo7e cells. The EC50 for this effect is 0.57-0.81 µg/mL (Routinely tested).