Learn More
ACROBiosystems ACROBiosystems Biotinylated Human IL-2 R beta / CD122 Protein, His,Avitag™
Catalog No. 501956504
Encompass_Preferred
Recombinant Protein;200UG;Biotinylated Human IL-2 R beta, His,Avitag (ILB-H82E3) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Asp 239 (Accession # P14784-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 28.2 kDa. The protein migrates as 30-32 kDa and 33-35 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-195-6504 Supplier ACROBiosystems Supplier No. ILBH82E3200UG
Recombinant Protein;200UG;Biotinylated Human IL-2 R beta, His,Avitag (ILB-H82E3) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Asp 239 (Accession # P14784-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 28.2 kDa. The protein migrates as 30-32 kDa and 33-35 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.