Learn More
ACROBiosystems ACROBiosystems Human IL-2 R gamma / CD132 Protein, Fc Tag
Catalog No. 501956848
Encompass_Preferred
Recombinant Protein;1MG;Human IL-2 R gamma, Fc Tag (ILG-H5256) is expressed from human 293 cells (HEK293). It contains AA Leu 23 - Asn 254 (Accession # AAH14972).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 53.5 kDa. The protein migrates as 75-96 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-195-6848 Supplier ACROBiosystems Supplier No. ILGH52561MG
Recombinant Protein;1MG;Human IL-2 R gamma, Fc Tag (ILG-H5256) is expressed from human 293 cells (HEK293). It contains AA Leu 23 - Asn 254 (Accession # AAH14972).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 53.5 kDa. The protein migrates as 75-96 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.