Learn More
ACROBiosystems ACROBiosystems Biotinylated Human IL-2 R gamma / CD132 Protein, His,Avitag™ (SPR verified)
Catalog No. 501956384
Encompass_Preferred
Recombinant Protein;25UG;Biotinylated Human IL-2 R gamma, His,Avitag (SPR verified) (ILG-H85E8) is expressed from Baculovirus-Insect cells (HI5). It contains AA Leu 23 - Asn 254 (Accession # P31785-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 31.0 kDa. The protein migrates as 40-50 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Biotinylated Human IL-2 R gamma, His,Avitag (SPR verified) (Cat. No. ILG-H85E8) immobilized on SA Chip can bind Human IL-2, Tag Free (Cat. No. IL2-H4113) with an affinity constant of 105 μM as determined in a SPR assay (Biacore T200) (QC tested).
Catalog No. 50-195-6384 Supplier ACROBiosystems Supplier No. ILGH85E825UG
Recombinant Protein;25UG;Biotinylated Human IL-2 R gamma, His,Avitag (SPR verified) (ILG-H85E8) is expressed from Baculovirus-Insect cells (HI5). It contains AA Leu 23 - Asn 254 (Accession # P31785-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 31.0 kDa. The protein migrates as 40-50 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Biotinylated Human IL-2 R gamma, His,Avitag (SPR verified) (Cat. No. ILG-H85E8) immobilized on SA Chip can bind Human IL-2, Tag Free (Cat. No. IL2-H4113) with an affinity constant of 105 μM as determined in a SPR assay (Biacore T200) (QC tested).