Learn More
AdipoGen IL-2 Superkine (Fc) (10 µg)
SDP
Supplier:  AdipoGen AG40B0111C010
Encompass_Preferred
Protein. The extracellular domain of human IL-2 (aa 21-153) (mutant H9 containing the mutations L80F / R81D / L85V / I 86V / I92F) is fused at the C-terminus to the Fc portion of human IgG2. Source: HEK 293 cells. Endotoxin content: <0.01EU/µg protein (LAL test; Lonza). Lyophilized. Contains PBS. Binds to human and mouse IL-2R. Purity: >95% (SDS-PAGE). IL-2 Superkine (Fc) is an artificial variant of IL-2 containing mutations at positions L80F / R81D / L85V / I 86V / I92F. These mutations are located in the molecule's core that acts to stabilize the structure and to give it a receptor-binding conformation mimicking native IL-2 bound to CD25. These mutations effectively eliminate the functional requirement of IL-2 for CD25 expression and elicit proliferation of T cells. Compared to IL-2, the IL-2 superkine induces superior expansion of cytotoxic T cells, leading to improved antitumour responses in vivo, and elicits proportionally less toxicity.
Catalog No. 50-205-3180