Learn More
Medchemexpress LLC IL-3 Mouse 10ug
SDP
Catalog No. 501934305
Encompass_Preferred
Recombinant mouse interleukin-3 (IL-3) is a lyophilized, biologically active cytokine that stimulates proliferation, differentiation, and survival of hematopoietic progenitors and myeloid-lineage cells. It is supplied as a high-purity, low-endotoxin preparation intended for in vitro research applications including cell culture, bioassays, and functional studies.
  • High biological activity (ED50 <0.2 ng/mL) suitable for sensitive assays.
  • Greater than 95% purity as determined by reducing SDS-PAGE.
  • Low endotoxin level (<0.2 EU/μg) compatible with cell-based experiments.
  • Lyophilized powder format enables long-term storage and convenient handling.
  • Recommended storage at -20°C; freeze aliquots for extended stability.
  • Includes the mouse IL-3 amino acid sequence (D33-C166); ~16 kDa by SDS-PAGE.
  • Do not reconstitute below 100 μg/mL; add carrier protein for long-term storage.
Catalog No. 50-193-4305 Supplier Medchemexpress LLC Supplier No. HYP706210UG
Recombinant mouse interleukin-3 (IL-3) is a lyophilized, biologically active cytokine that stimulates proliferation, differentiation, and survival of hematopoietic progenitors and myeloid-lineage cells. It is supplied as a high-purity, low-endotoxin preparation intended for in vitro research applications including cell culture, bioassays, and functional studies.
  • High biological activity (ED50 <0.2 ng/mL) suitable for sensitive assays.
  • Greater than 95% purity as determined by reducing SDS-PAGE.
  • Low endotoxin level (<0.2 EU/μg) compatible with cell-based experiments.
  • Lyophilized powder format enables long-term storage and convenient handling.
  • Recommended storage at -20°C; freeze aliquots for extended stability.
  • Includes the mouse IL-3 amino acid sequence (D33-C166); ~16 kDa by SDS-PAGE.
  • Do not reconstitute below 100 μg/mL; add carrier protein for long-term storage.