Learn More
Sigma Aldrich Fine Chemicals Biosciences IL 3 human Animal componen
Catalog No. 502905029
Encompass
IL3 (interleukin 3) is a haematopoietic growth factor the gene for which is localized to human chromosome locus 5. Recombinant human IL-3 is a 15.0 kDa globular protein containing 133 amino acid residues.
Catalog No. 50-290-5029 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. SRP309010UG
IL3 (interleukin 3) is a haematopoietic growth factor the gene for which is localized to human chromosome locus 5. Recombinant human IL-3 is a 15.0 kDa globular protein containing 133 amino acid residues.