Learn More
Enzo Life Sciences IL-3 (mouse), (recombinant) (10µg)
SDP
Supplier:  Enzo Life Sciences ALX2018210010
Encompass_Preferred
Alternative name: Interleukin-3, MCGF, Multi-CSF, HCGF, P-cell stimulation factor. Endotoxin: ≤1 EU/μg protein measured by kinetic LAL analysis. Purity: ≥95% (reducing and non-reducing SDS-PAGE). MW: ~15.1kDa. Appearance: White lyophilized (freeze-dried) powder. Formulation: Lyophilized. Sterile filtered. Source: Produced in E. coli. Non-glycosylated protein, containing 135 amino acids. Long Term Storage: -20°C. Use/Stability: It is recommended that a carrier protein (0.1% HSA or BSA) is added for long term storage. Handling: Centrifuge the vial before opening the cap. After reconstitution, prepare aliquots and store at -20°C.
Catalog No. 50-200-8029