Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Interleukin-31 (canine) | >95.0% | 20UG
SDP

Supplier:  Medchemexpress LLC HYP70371820UG

Encompass_Preferred

Recombinant canine interleukin-31 expressed in HEK293 mammalian cells and supplied as a lyophilized N-His-tagged protein for research use. The product is provided as a stabilized lyophilized powder formulated in PBS with trehalose, exhibits glycosylation-dependent mobility on SDS-PAGE, and should be stored frozen and handled per supplier recommendations to preserve activity.

  • Produced in HEK293 mammalian cells for native-like glycosylation.
  • Contains N-terminal His tag to enable purification and detection.
  • Lyophilized powder stabilized with trehalose for storage and shipping.
  • Low endotoxin level (<1 EU/μg) suitable for sensitive assays.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Molecular weight approximately 26-36 kDa due to glycosylation.
  • Reconstitute and store according to supplier recommendations to preserve activity.
  • For research use only; not for clinical or diagnostic applications.

Catalog No. 50-001-75249


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.