Learn More
Medchemexpress LLC IL-31 Cynomolgus H 10ug | 10ug
SDP
Catalog No. 5000186193
Encompass_Preferred
IL-31 is a multifunctional cytokine that activates STAT3 through IL-31 heterodimeric receptors (IL31RA and OSMR) and may activate STAT1 and STAT5 IL-31 is known for its involvement in skin immunity where it regulates immune responses IL-31 Protein Cynomolgus (HEK293 His) is the recombinant cynomolgus-derived IL-31 protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-86193 Supplier Medchemexpress LLC Supplier No. HYP7700610UG
IL-31 is a multifunctional cytokine that activates STAT3 through IL-31 heterodimeric receptors (IL31RA and OSMR) and may activate STAT1 and STAT5 IL-31 is known for its involvement in skin immunity where it regulates immune responses IL-31 Protein Cynomolgus (HEK293 His) is the recombinant cynomolgus-derived IL-31 protein expressed by HEK293 with C-His labeled tag