Learn More
Medchemexpress LLC IL-31 Human HEK293 50ug
SDP
Catalog No. 5000181243
Encompass_Preferred
IL-31 is a multifunctional cytokine that activates STAT3 through IL-31 heterodimeric receptors (IL31RA and OSMR) and may activate STAT1 and STAT5 IL-31 is known for its involvement in skin immunity where it regulates immune responses IL-31 Protein Human (HEK293 His) is the recombinant human-derived IL-31 protein expressed by HEK293 with C-10 His labeled tag
Catalog No. 50-001-81243 Supplier Medchemexpress LLC Supplier No. HYP7320950UG
IL-31 is a multifunctional cytokine that activates STAT3 through IL-31 heterodimeric receptors (IL31RA and OSMR) and may activate STAT1 and STAT5 IL-31 is known for its involvement in skin immunity where it regulates immune responses IL-31 Protein Human (HEK293 His) is the recombinant human-derived IL-31 protein expressed by HEK293 with C-10 His labeled tag