Learn More
Medchemexpress LLC IL-31 protein, mouse (HEK293, His) | >85.0% | 100 UG
SDP
Catalog No. 5000291232
Encompass_Preferred
Recombinant mouse interleukin-31 (IL-31) expressed in HEK293 cells and supplied as a 0.22 μm filtered PBS solution. The protein carries a C-terminal His tag, is >85% pure by reducing SDS-PAGE, and shows functional activity in receptor binding ELISA (ED50 = 2.916 μg/mL). It is intended for in vitro signaling, receptor binding, and assay development.
Catalog No. 50-002-91232 Supplier Medchemexpress LLC Supplier No. HYP74804100UG
Recombinant mouse interleukin-31 (IL-31) expressed in HEK293 cells and supplied as a 0.22 μm filtered PBS solution. The protein carries a C-terminal His tag, is >85% pure by reducing SDS-PAGE, and shows functional activity in receptor binding ELISA (ED50 = 2.916 μg/mL). It is intended for in vitro signaling, receptor binding, and assay development.