Learn More
ACROBiosystems ACROBiosystems Human IL-31 RA Protein, Fc Tag
Catalog No. 501956906
Encompass_Preferred
Recombinant Protein;1MG;Human IL-31 RA, Fc Tag (ILA-H5256) is expressed from human 293 cells (HEK293). It contains AA Ala 20 - Glu 519 (Accession # Q8NI17-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 83.5 kDa. The protein migrates as 105-115 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-195-6906 Supplier ACROBiosystems Supplier No. ILAH52561MG
Recombinant Protein;1MG;Human IL-31 RA, Fc Tag (ILA-H5256) is expressed from human 293 cells (HEK293). It contains AA Ala 20 - Glu 519 (Accession # Q8NI17-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 83.5 kDa. The protein migrates as 105-115 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.