Learn More
Medchemexpress LLC IL-32 Human HEK293 1mg
SDP
Catalog No. 5000179406
Encompass_Preferred
IL-32 protein is a member of the cytokine family whose expression increases following T cell activation or IL-2-induced NK cell activation It plays a key role in inducing macrophages to produce TNF which contributes to the inflammatory response IL-32 alphaProtein Human (HEK293 His) is the recombinant human-derived IL-32 alpha protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-79406 Supplier Medchemexpress LLC Supplier No. HYP738841MG
IL-32 protein is a member of the cytokine family whose expression increases following T cell activation or IL-2-induced NK cell activation It plays a key role in inducing macrophages to produce TNF which contributes to the inflammatory response IL-32 alphaProtein Human (HEK293 His) is the recombinant human-derived IL-32 alpha protein expressed by HEK293 with C-His labeled tag