Learn More
ACROBiosystems ACROBiosystems Human IL-33 Protein, His Tag
Catalog No. 501956842
Encompass_Preferred
Recombinant Protein;1MG;Human IL-33, His Tag (IL3-H52H7) is expressed from human 293 cells (HEK293). It contains AA His 109 - Thr 270 (Accession # O95760-1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 20.2 kDa. The protein migrates as 25-30 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human IL-1RL1, Fc Tag (Cat. No. IL1-H5250) at 5 μg/mL (100 μL/well) can bind Human IL-33, His Tag (Cat. No. IL3-H52H7) with a linear range of 5-20 ng/mL (QC tested).
Catalog No. 50-195-6842 Supplier ACROBiosystems Supplier No. IL3H52H71MG
Recombinant Protein;1MG;Human IL-33, His Tag (IL3-H52H7) is expressed from human 293 cells (HEK293). It contains AA His 109 - Thr 270 (Accession # O95760-1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 20.2 kDa. The protein migrates as 25-30 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human IL-1RL1, Fc Tag (Cat. No. IL1-H5250) at 5 μg/mL (100 μL/well) can bind Human IL-33, His Tag (Cat. No. IL3-H52H7) with a linear range of 5-20 ng/mL (QC tested).