Learn More
Enzo Life Sciences IL-33 (human), (recombinant) (10µg)
SDP
Supplier:  Enzo Life Sciences ALX5220983010
Encompass_Preferred
Alternative name: Interleukin-33, NF-HEV, IL-1F11. Concentration: 0.1mg/ml after reconstitution. Purity: ≥95% (SDS-PAGE). Formulation: Lyophilized. Contains PBS. Source: Produced in E. coli. Untagged human IL-33 (aa 112-270). Specificity: Binds to human ST2. Long Term Storage: -20°C.
Catalog No. 50-200-9416