Learn More
AdipoGen IL-33 human pAb
SDP
Supplier:  AdipoGen AG25A0045C100
Encompass_Preferred
Polyclonal Antibody. Recognizes human IL-33. Detects a band of ~18kDa by Western blot. Source Rabbit. Applications ELISA, IHC, WB. Liquid. 0.2µm-filtered solution in PBS, pH 7.4. Contains no preservatives. Interleukin-33 IL-33 HF-NEV IL-1F11, a member of the IL-1 family of cytokines, is expressed by many cell types following pro-inflammatory stimulation and is thought to be released on cell lysis. The 30kDa human IL33 is converted by CASP1 to a 18kDa protein. IL33 binds to and signals through ST2 IL1R1 and its stimulation recruits MYD88, IRAK, IRAK4, and TRAF6, followed by phosphorylation of ERK1MAPK3/ERK2MAPK1, p38MAPK14, and JNK. The ability of IL-33 to target numerous immune cell types, like Th2-like cells, mast cells, and B1 cells, and to induce cytokine and chemokine production underlines its potential in influencing the outcome of a wide range of diseases, such as arthritis, asthma, atopic allergy & anaphylaxis, cardiovascular disease/atherosclerosis.
Catalog No. 50-256-3267