Learn More
AdipoGen anti-IL-33, mAb (IL33026B) (50 µg)
SDP
Supplier:  AdipoGen AG20A0043C050
Encompass_Preferred
Monoclonal Antibody. Recognizes human and mouse IL-33. Isotype: Mouse IgG1kappa. Clone: IL33026B. Applications: ELISA, IP, WB. Liquid. 0.2µm-filtered solution in PBS, pH 7.4. Interleukin-33 (IL-33; HF-NEV; IL-1F11), a member of the IL-1 family of cytokines, is expressed by many cell types following pro-inflammatory stimulation and is thought to be released on cell lysis. The 30kDa human IL33 is converted by CASP1 to a 18kDa protein. IL33 binds to and signals through ST2 (IL1R1) and its stimulation recruits MYD88, IRAK, IRAK4, and TRAF6, followed by phosphorylation of ERK1(MAPK3)/ERK2(MAPK1), p38(MAPK14), and JNK. The ability of IL-33 to target numerous immune cell types, like Th2-like cells, mast cells, and B1 cells, and to induce cytokine and chemokine production underlines its potential in influencing the outcome of a wide range of diseases, such as arthritis, asthma, atopic allergy & anaphylaxis, cardiovascular disease/atherosclerosis, nervous system diseases, and sepsis.
Catalog No. 50-205-2838